missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB2 Antibody (SAIC-19D-1F4) - Chimeric - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-09000-0.025mg
This item is not returnable.
View return policy
Description
GRB2 Monoclonal antibody specifically detects GRB2 in Human samples. It is validated for Western Blot, Mass Spectrometry
Specifications
| GRB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Mass Spectrometry | |
| SAIC-19D-1F4 | |
| Unconjugated | |
| abundant SRC homology, Adapter protein GRB2, ASH, EGFRBP-GRB2, epidermal growth factor receptor-binding protein GRB2, Grb3-3, growth factor receptor-bound protein 2, growth factor receptor-bound protein 3, HT027, MST084, MSTP084, NCKAP2, Protein Ash, SH2/SH3 adapter GRB2 | |
| Peptide 'FGNDVQHFK', derived from the Growth factor receptor-bound protein 2, conjugated to KLH. | |
| 0.025 mg | |
| Angiogenesis, Breast Cancer, Cancer, Extracellular Matrix, Signal Transduction | |
| 2885 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction