missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB7 Antibody (OTI6H2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00 € - 550.00 €
Specifications
| Antigen | GRB7 |
|---|---|
| Clone | OTI6H2 |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:2000 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306677
|
Novus Biologicals
NBP3-44930-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306272
|
Novus Biologicals
NBP3-44930-30ul |
30 μL |
256.00 €
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRB7 Monoclonal antibody specifically detects GRB7 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| GRB7 | |
| Western Blot 1:2000, Immunohistochemistry 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| B47, Epidermal growth factor receptor GRB-7, GRB7 adapter protein, growth factor receptor-bound protein 7 | |
| Full length human recombinant protein of human GRB7 (NP_005301) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI6H2 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Extracellular Matrix | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 2886 | |
| IgG2b | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title