missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRB7 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148920
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with epidermal growth factor receptor (EGFR) and ephrin receptors. The protein plays a role in the integrin signaling pathway and cell migration by binding with focal adhesion kinase (FAK). Alternative splicing results in multiple transcript variants encoding different isoforms, although the full-length natures of only two of the variants have been determined to date.
Specifications
| GRB7 | |
| Polyclonal | |
| Unconjugated | |
| GRB7 | |
| B47; Epidermal growth factor receptor GRB-7; Grb7; GRB7 adapter protein; growth factor receptor binding protein GRB7; growth factor receptor bound protein 7; growth factor receptor-bound protein 7; Growth factor receptor-bound protein 7 (GRB7 adapter protein) (Epidermal growth factor receptor GRB-7); mKIAA4028; OTTHUMP00000164352 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 14786, 2886, 84427 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q03160, Q14451, Q9QZC5 | |
| GRB7 | |
| E. coli-derived recombinant human GRB7, Met130-Tyr274. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction