Learn More
GREM2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | GREM2 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18078985
|
Proteintech
CL488-13892-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095205
|
Proteintech
CL488-13892-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GREM2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GREM2. This antibody recognizes human, mouse antigen. The GREM2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| GREM2 | |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse | |
| CKTSF1B2, DAN domain family member 3, DAND3, GREM2, Gremlin 2, PRDC | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 64388 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.