missing translation for 'onlineSavingsMsg'
Learn More

GRHL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

113.00 € - 363.00 €

Specifications

Antigen GRHL1
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18696024
View Documents
Novus Biologicals
NBP3-21433-100ul
100 μg
363.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18680674
View Documents
Novus Biologicals
NBP3-21433-25ul
25 μg
113.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GRHL1 Polyclonal antibody specifically detects GRHL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

GRHL1
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
grainyhead-like 1 (Drosophila), grainyhead-like protein 1 homolog, LBP protein 32, MGRLBP-32, NH32, TFCP2L2, Transcription factor CP2-like 2LBP32Mammalian grainyhead, Transcription factor LBP-32
Produced in rabbits immunized with E. coli-derived Human GRHL1 fragment
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000
Polyclonal
Purified
RUO
PBS, pH 7.0
29841
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.