missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRHL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
378.00 € - 572.00 €
Specifications
| Antigen | GRHL1 |
|---|---|
| Applications | ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18468340
|
Novus Biologicals
NBP1-81321-25ul |
25ul |
378.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18051964
|
Novus Biologicals
NBP1-81321 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRHL1 Polyclonal specifically detects GRHL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).Specifications
| GRHL1 | |
| Polyclonal | |
| Rabbit | |
| Rat, Human, Mouse | |
| grainyhead-like 1 (Drosophila), grainyhead-like protein 1 homolog, LBP protein 32, MGRLBP-32, NH32, TFCP2L2, Transcription factor CP2-like 2LBP32Mammalian grainyhead, Transcription factor LBP-32 | |
| GRHL1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human GRHL1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 29841 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TVFKPFIDLDTQPVLFIPDVHFANLQRGTHVLPIASEELEGEGSVLKRGPYGTEDDFAVPPSTKLARIEEPKRVLLYVRKESEEVFDALMLKTPSLKGL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title