Learn More
Gilac™ Grid
Product Code. 17928711
Change view
Click to view available options
For Use With (Equipment):
10 L Flat Tray
2 L Flat Tray
3 L Flat Tray
5 L Flat Tray
600 x 400 Container
8 L Flat Tray
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17928711 600 x 400 Container Each
17958781 2 L Flat Tray Each
17958421 3 L Flat Tray Each
17988701 5 L Flat Tray Each
17978701 8 L Flat Tray Each
17948561 10 L Flat Tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17928711 Supplier Gilac™ Supplier No. G100421
  • ONLINE EXCLUSIVE PRICE
    9.06 €
    5.99 € / Pack of 1
    Save 3.07 €
    33% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Grid is highly durable and shock resistant; it avoids any contact between food and exudates.
  • Grid for flat trays.
  • Highly durable and shock resistant.
  • Enables the use of flat dishes as defrosting trays by separating the ice water from the product being defrosted.
Specifications
Product Type Container Grid
Material Polyethylene
For Use With (Equipment) 600 x 400 Container