Learn More
GSK-3 alpha/beta Antibody (S08-8G7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19938-100ul
 View more versions of this product
Product Code. 18613335
  • 645.00 € / 100µL
Description
GSK-3 alpha/beta Monoclonal antibody specifically detects GSK-3 alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GSK-3 alpha/beta
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S08-8G7
Unconjugated
DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha
A synthetic peptide of human GSK-3 alpha/beta (Uniprot # P49840)
100 μg
Cell Biology, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers
2931
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.