missing translation for 'onlineSavingsMsg'
Learn More

GSK137647A, MedChemExpress

Product Code. 16468008
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468008 10 mg 10mg
16457786 100 mg 100mg
16423937 50 mg 50mg
16423736 25 mg 25mg
16400478 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16468008 Supplier Medchem Express Supplier No. HY19995
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK137647A (GSK 137647) is a potent, selective free fatty acid receptor 4 (FFA4) agonist with pEC50 values of 6.3, 6.2, and 6.1 for human, mouse and rat FFA4, and pEC50 values < 4.5 for all three species for FFA1, FFA2, and FFA3, respectively.

GSK137647A has anti-inflammatory activity. GSK137647A induces insulin secretion and inhibits epithelial ion transport, GSK137647A is related to regulation of glucose homeostasis and anti-inflammatory response.

Specifications

Chemical Name or Material GSK137647A
CAS 349085-82-1
Color White
Purity Grade Notes Research
Molecular Formula C16H19NO3S
Quantity 10 mg
Synonym GSK 137647
Solubility Information DMSO : ≥ 100 mg/mL (327.45 mM) ∣H2O : mg/mL (ultrasonic;warming) (Need ultrasonic and warming)
SMILES O=S(C1=CC=C(OC)C=C1)(NC2=C(C)C=C(C)C=C2C)=O
Molecular Weight (g/mol) 305.39
Formula Weight 305.39
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.