missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK180736A, MedChemExpress
missing translation for 'orderingAttributeHoverText'
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
2mg
50mg
5mg
Description
GSK180736A is potent Rho-associated coiled-coil kinase 1 (ROCK1) inhibitor with an IC50 of 100 nM. GSK180736A is also a selective and ATP-competitive G protein-coupled receptor kinase 2 (GRK2) inhibitor with an IC50 of 0.77 μM.
Specifications
Specifications
| Chemical Name or Material | GSK180736A |
| CAS | 817194-38-0 |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C19H16FN5O2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 30 mg/mL (82.11 mM) |
| SMILES | CC(N1)=C(C(NC2=CC=C(NN=C3)C3=C2)=O)C(C4=CC=C(F)C=C4)NC1=O |
| Molecular Weight (g/mol) | 365.36 |
| Formula Weight | 365.36 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction