missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK481, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Description
GSK481 is a highly potent, selective, and specific receptor interacting protein 1 (RIP1) kinase inhibitor with an IC50 of 1.3 nM, which inhibits Ser166 phosphorylation in wild-type human RIP1 (IC50=2.8 nM). GSK481 also exhibits excellent translation in the U937 cellular assay with an IC50 of 10 nM.
Specifications
Specifications
| Chemical Name or Material | GSK481 |
| CAS | 1622849-58-4 |
| Color | Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C21H19N3O4 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 35 mg/mL (92.74 mM) |
| SMILES | O=C(C1=NOC(CC2=CC=CC=C2)=C1)N[C@H]3COC4=CC=CC=C4N(C)C3=O |
| Molecular Weight (g/mol) | 377.39 |
| Formula Weight | 377.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction