missing translation for 'onlineSavingsMsg'
Learn More

GSK598809, MedChemExpress

Product Code. 16483787
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483787 50 mg 50mg
16467838 10 mg 10mg
16427666 100 mg 100mg
16410308 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483787 Supplier Medchem Express Supplier No. HY19654
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK598809 is a potent and selective dopamine D3 Receptor (DRD3) antagonist, with a pKi of 8.9.

GSK598809 is a potent and selective dopamine D3 Receptor (DRD3) antagonist, with a pKi of 8.9.

Specifications

Chemical Name or Material GSK598809
CAS 863680-45-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H23F4N5OS
Quantity 50 mg
Solubility Information DMSO : 83.33 mg/mL (173.06 mM; Need ultrasonic) ∣1M HCl : 28.57 mg/mL (59.33 mM; ultrasonic and adjust pH to 2 with HCl)
SMILES CC1=C(C2=NN=C(SCCCN3CC4(C5=CC=C(C(F)(F)F)C=C5F)CC4C3)N2C)OC=N1
Molecular Weight (g/mol) 481.51
Formula Weight 481.51
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.