missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSPT1 Antibody (CL13332), Novus Biologicals™
Mouse Monoclonal Antibody
437.00 € - 693.00 €
Specifications
| Antigen | GSPT1 |
|---|---|
| Clone | CL13332 |
| Dilution | Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673575
|
Novus Biologicals
NBP3-18565-100ul |
100 μg |
693.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660485
|
Novus Biologicals
NBP3-18565-25ul |
25 μg |
437.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSPT1 Monoclonal antibody specifically detects GSPT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| GSPT1 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ERF3A, eRF3aFLJ38048, ETF3A, eukaryotic peptide chain release factor GTP-binding subunit ERF3A, Eukaryotic peptide chain release factor subunit 3a, FLJ39067, G1 to S phase transition 1,551G9.2, G1 to S phase transition protein 1 homolog, GST1 | |
| This antibody was developed against a sythetic peptide for P15170. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL13332 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| PBS, pH 7.2, 40% glycerol | |
| 2935 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title