missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSPT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 585.00 €
Specifications
| Antigen | GSPT1 |
|---|---|
| Applications | KnockDown, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18013696
|
Novus Biologicals
NBP2-47442 |
0.1 mL |
585.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642816
|
Novus Biologicals
NBP2-47442-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSPT1 Polyclonal specifically detects GSPT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| GSPT1 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 2935 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GGAANNHGAGSGAGGRAAPVESSQEEQSLCEGSNSAVS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| KnockDown, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| ERF3A, eRF3aFLJ38048, ETF3A, eukaryotic peptide chain release factor GTP-binding subunit ERF3A, Eukaryotic peptide chain release factor subunit 3a, FLJ39067, G1 to S phase transition 1,551G9.2, G1 to S phase transition protein 1 homolog, GST1 | |
| GSPT1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title