missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-GST (Rabbit) Antibody
Rabbit Polyclonal Antibody
1192.00 € - 14300.00 €
Specifications
| Antigen | GST |
|---|---|
| Host Species | Rabbit |
| Applications | Kinase Assay |
| Regulatory Status | RUO |
| Conjugate | Tb (Terbium) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15255684
|
Thermo Scientific™
A15112 |
25 μg |
1192.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15265684
|
Thermo Scientific™
A15113 |
100 μg |
3015.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15275684
|
Thermo Scientific™
A15114 |
1 mg |
14300.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- LanthaScreen™ terbium-labeled anti-GST Rabbit antibodies are used to indirectly label proteins tagged with GST
- This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity
Specifications
| GST | |
| Kinase Assay | |
| Tb (Terbium) | |
| Terbium-labeled anti-GST antibody in HEPES buffered saline. Store at -20°C. | |
| Polyclonal |
| Rabbit | |
| RUO | |
| Frozen | |
| Primary | |
| in HEPES buffered Saline |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title