missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTA4 Antibody (SR1360), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22646-50ul
This item is not returnable.
View return policy
Description
GSTA4 Monoclonal antibody specifically detects GSTA4 in Human, Rat samples. It is validated for Western Blot
Specifications
| GSTA4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| SR1360 | |
| Western Blot 1:500-1:2000 | |
| DKFZp686D21185, EC 2.5.1.18, glutathione S-alkyltransferase A4, glutathione S-aralkyltransferase A4, glutathione S-aryltransferase A4, glutathione S-transferase A4, Glutathione S-transferase A4-4, glutathione S-transferase alpha 4, glutathione transferase A4-4, GST class-alpha member 4, GSTA4-4, GTA4, S-(hydroxyalkyl)glutathione lyase A4 | |
| A synthesized peptide derived from human GSTA4 (Uniprot #: O15217) | |
| 50 μg | |
| metabolism | |
| 2941 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction