missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTP1 Recombinant Rabbit Monoclonal Antibody (23GB6420), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | GSTP1 |
|---|---|
| Clone | 23GB6420 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088459
![]() |
Invitrogen™
MA553958 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089035
|
Invitrogen™
MA553959 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSTP1 Recombinant Monoclonal antibody specifically detects GSTP1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GSTP1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P04906, P09211, P19157 | |
| 14870, 24426, 2950 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6420 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Chain 7; deafness, X-linked 7; DFN7; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-transferase P; Glutathione S-transferase P 1; glutathione S-transferase pi 1; glutathione S-transferase pi 2; glutathione S-transferase, pi 1; glutathione S-transferase, pi 2; Glutathione-S-transferase placental enzyme pi type; glutathione-S-transferase, pi 1; Glutathione-S-transferase, placental enzyme pi type; GST 7-7; GST class-pi; gst P1; Gst p-1; GST YF-YF; Gst3; Gstp; GST-P; Gstp1; GSTP1-1; Gstp2; GstpiB; GST-piB; HEL-S-22; HGNC:4638; PI; Preadipocyte growth factor | |
| A synthesized peptide derived from human GST3/GST pi (100-201AA). | |
| Primary | |
| -20°C | |
| Gstp1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title