missing translation for 'onlineSavingsMsg'
Learn More

GSTZ1 Monoclonal Antibody (OTI1A7), TrueMAB™, OriGene

Product Code. 17523914
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Conditionnement:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
17523914 30 μL 30µL
17503914 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17523914 Fournisseur Origene Technologies Code fournisseur TA810615S

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

GSTZ1 Monoclonal antibody specifically detects GSTZ1 in Human samples. It is validated for Western Blot

Specifica

Antigen GSTZ1
Applications Western Blot
Classification Monoclonal
Clone OTI1A7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Mme
Gene Accession No. P08473
Gene Alias 17-beta-HSD 10; 17-beta-hydroxysteroid dehydrogenase 10; 17b-HSD10; 2-methyl-3-hydroxybutyryl-CoA dehydrogenase; 3-hydroxy-2-methylbutyryl-CoA dehydrogenase; 3-hydroxyacyl-CoA dehydrogenase type II; 3-hydroxyacyl-CoA dehydrogenase type-2; ABAD; AB-binding alcohol dehydrogenase; amyloid-beta peptide binding alcohol dehydrogenase; CAMR; DUPXp11.22; Endoplasmic reticulum-associated amyloid beta-peptide-binding protein; ERAB; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase alpha 3; glutathione S-transferase zeta 1; glutathione transferase zeta 1; glutathione transferase zeta 1 (maleylacetoacetate isomerase); GSTZ1; GSTZ1-1; HADH2; HCD2; HSD17B10; hydroxysteroid (17-beta) dehydrogenase 10; hydroxysteroid 17-beta dehydrogenase 10; hydroxysteroid dehydrogenase 10; MAAI; MAI; Maleylacetoacetate isomerase; maleylacetone isomerase; MHBD; Mitochondrial ribonuclease P protein 2; Mitochondrial RNase P protein 2; mitochondrial RNase P subunit 2; MRPP2; MRX17; MRX31; MRXS10; S-(hydroxyalkyl)glutathione lyase; SCHAD; SDR5C1; Short chain dehydrogenase/reductase family 5C member 1; short chain dehydrogenase/reductase family 5C, member 1; short chain L-3-hydroxyacyl-CoA dehydrogenase type 2; short chain type dehydrogenase/reductase XH98G2; Short-chain type dehydrogenase/reductase XH98G2; Type II HADH; XH98G2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 411-750 of human MME produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4311
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.