missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2A1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97826-50ul
This item is not returnable.
View return policy
Description
GTF2A1L Polyclonal antibody specifically detects GTF2A1L in Human samples. It is validated for Western Blot
Specifications
| GTF2A1L | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 1-like factor, general transcription factor IIA, 1-like, GTF2A1LF, MGC26254, TFIIA large subunit isoform ALF, TFIIA-alpha and beta-like factor, TFIIA-alpha/beta-like factor | |
| Produced in rabbits immunized with E. coli-derived Human GTF2A1L fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 11036 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction