missing translation for 'onlineSavingsMsg'
Learn More

GTF2A1L Monoclonal Antibody (OTI4B4), TrueMAB™, OriGene

Product Code. 17524514
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17524514 30 μL 30µL
17504514 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17524514 Supplier Origene Technologies Supplier No. TA810961S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GTF2A1L Monoclonal antibody specifically detects GTF2A1L in Human samples. It is validated for Western Blot

Specifications

Antigen GTF2A1L
Applications Western Blot
Classification Monoclonal
Clone OTI4B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GTF2A1L
Gene Accession No. Q9UNN4
Gene Alias ALF; CXCDC1; general transcription factor II A, 1-like factor; general transcription factor IIA 1 like; general transcription factor IIA 1-like; general transcription factor IIA subunit 1 like; general transcription factor IIA, 1-like; GTF2A1L; GTF2A1LF; GTF2A1-like factor; KIAA2037; testis secretory sperm-binding protein Li 230m; TFIIA large subunit isoform ALF; TFIIA-alpha and beta-like factor; TFIIA-alpha/beta-like factor
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 183-478 of human GTF2A1L produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11036
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.