missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TFIIF is a general transcription initiation factor that binds to RNA polymerase II and helps to recruit it to the initiation complex in collaboration with TFIIB. It promotes transcription elongation.
Specifications
Specifications
| Antigen | GTF2F1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.28 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | GTF2F1 |
| Gene Accession No. | P35269, Q3THK3, Q6AY96 |
| Gene Alias | BTF4, GTF2F1, RAP74, TF2F1, TFIIF, TFIIF alpha |
| Gene Symbols | GTF2F1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?