missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38556
This item is not returnable.
View return policy
Description
GTF2H1 Polyclonal specifically detects GTF2H1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GTF2H1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| P32780 | |
| GTF2H1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PEGKAKIQLQLVLHAGDTTNFHFSNESTAVKERDAVKDLLQQLLPKFKRKANKELEEKNRMLQEDPVLFQLYKDLVVSQVISAEEFWANRLNVNATDSSSTSNHKQDVGISAAFLADVRPQTDGCNGLRYN | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair | |
| 2965 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Basic transcription factor 2 62 kDa subunit, BTF2 p62, BTF2general transcription factor IIH, polypeptide 1 (62kD subunit), General transcription factor IIH polypeptide 1, general transcription factor IIH subunit 1, general transcription factor IIH, polypeptide 1, 62kDa, TFB1, TFIIH, TFIIH basal transcription factor complex p62 subunit | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction