missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTP binding protein era homolog Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89704
This item is not returnable.
View return policy
Beschreibung
GTP binding protein era homolog Polyclonal specifically detects GTP binding protein era homolog in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
| GTP binding protein era homolog | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20 | |
| CEGA, Conserved ERA-like GTPase, ERA, Era (E. coli G-protein homolog)-like 1, Era G-protein-like 1 (E. coli), ERAL1A, ERA-like protein 1, ERA-W, GTPase Era, mitochondrial, GTPase, human homolog of E. coli essential cell cycle protein Era, GTP-binding protein era homolog, HERA, H-ERA, HERA-A, HERA-B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26284 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ERAL1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PWEYHSAVLTSQTPEEICANIIREKLLEHLPQEVPYNVQQKTAVWEEGPGGELVIQQKLLVPKESYVKLLIGPKGHVISQIAQEAGHDLMDIFLCDVDIRLSVKLLK | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur