missing translation for 'onlineSavingsMsg'
Learn More

Guanethidine sulfate, MedChemExpress

Product Code. 16438466
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438466 100 mg 100mg
16432216 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438466 Supplier Medchem Express Supplier No. HYB0800
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Guanethidine sulfate (Guanethidine monosulfate) ia an antihypertensive agents. Guanethidine is also an adrenergic neurone blocking drug, enters noradrenergic nerve terminals by the neuronal amine carrier.

Guanethidine sulfate (Guanethidine monosulfate) ia an antihypertensive agents. Guanethidine is also an adrenergic neurone blocking drug, enters noradrenergic nerve terminals by the neuronal amine carrier.

Specifications

Chemical Name or Material Guanethidine sulfate
Physical Form Solid
CAS 645-43-2
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H24N4O4S
For Use With (Application) Neuroscience-Neuromodulation
Synonym Guanethidine monosulfate
Solubility Information H2O : 100 mg/mL (337.39 mM; Need ultrasonic)
SMILES NC(NCCN1CCCCCCC1)=N.O=S(O)(O)=O
Molecular Weight (g/mol) 296.39
Formula Weight 296.39
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.