missing translation for 'onlineSavingsMsg'
Learn More

Guanfacine hydrochloride, MedChemExpress

Product Code. 16477328
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477328 10 mg 10mg
16447296 100 mg 100mg
16443347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16477328 Supplier Medchem Express Supplier No. HY17416
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Guanfacine hydrochloride, an anti-hypertensive agent, is a selective α2A-adrenoceptor agonist with Kd of 31 nM and displays 60-fold selectivity over α2B-adrenoceptors.

Guanfacine hydrochloride, an anti-hypertensive agent, is a selective α2A-adrenoceptor agonist with Kd of 31 nM and displays 60-fold selectivity over α2B-adrenoceptors.

Specifications

Chemical Name or Material Guanfacine hydrochloride
Physical Form Solid
CAS 29110-48-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C9H10Cl3N3O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 30 mg/mL (106.18 mM) ∣H2O : 20 mg/mL (70.78 mM; Need ultrasonic)
SMILES O=C(NC(N)=N)CC1=C(Cl)C=CC=C1Cl.[H]Cl
Molecular Weight (g/mol) 282.55
Formula Weight 282.55
Percent Purity 99.81%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.