missing translation for 'onlineSavingsMsg'
Learn More

Guanfacine Hydrochloride, TRC

Product Code. 30240810
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240810 10 mg 10mg
30240597 25 mg 25mg
30240728 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240810 Supplier Toronto Research Chemicals Supplier No. TRCG81600110MG
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29110-48-3
Molecular Weight (g/mol) 282.55
Synonym N-(Aminoiminomethyl)-2,6-dichloro-benzeneacetamide Hydrochloride (1:1), Benzeneacetamide, N-(aminoiminomethyl)-2,6-dichloro-, monohydrochloride (9CI), 2,6-Dichlorophenylacetylguanidine hydrochloride, BS 100-141, Estulic, Guanfacine hydrochloride, INTUNIV, LON 798, N-Amidino-2-(2,6-dichlorophenyl)acetamide hydrochloride, SPD 503, Tenex, Tenex (pharmaceutical), [(2,6-Dichlorophenyl)acetyl]guanidine monohydrochloride
IUPAC Name N-carbamimidoyl-2-(2,6-dichlorophenyl)acetamide;hydrochloride
SMILES Cl.NC(=N)NC(=O)Cc1c(Cl)cccc1Cl

Specifications

Molecular Formula C9 H9 Cl2 N3 O . Cl H
Quantity 10 mg
InChI Formula InChI=1S/C9H9Cl2N3O.ClH/c10-6-2-1-3-7(11)5(6)4-8(15)14-9(12)13;/h1-3H,4H2,(H4,12,13,14,15);1H
Formula Weight 280.9889
Chemical Name or Material Guanfacine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.