missing translation for 'onlineSavingsMsg'
Learn More

GW 501516, MedChemExpress

Product Code. 16494188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494188 10 mg 10mg
16480537 50 mg 50mg
16476907 5 mg 5mg
16425156 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494188 Supplier Medchem Express Supplier No. HY10838
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GW 501516 (GW 1516) is a PPARδ agonist with an EC50 of 1.1 nM.

GW 501516 (GW 1516) is a PPARδ agonist with an EC50 of 1.1 nM.

Specifications

Chemical Name or Material GW 501516
CAS 317318-70-0
Color White
Purity Grade Notes Research
Molecular Formula C21H18F3NO3S2
Quantity 10 mg
Synonym GW 1516 GSK-516
Solubility Information DMSO : ≥ 100 mg/mL (220.51 mM)
SMILES OC(COC1=CC=C(C=C1C)SCC2=C(N=C(S2)C3=CC=C(C=C3)C(F)(F)F)C)=O
Molecular Weight (g/mol) 453.5
Formula Weight 453.5
Percent Purity 97.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.