missing translation for 'onlineSavingsMsg'
Learn More

GW 501516 Methyl Ester, TRC

Product Code. 30009154
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009154 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30009154 Supplier Toronto Research Chemicals Supplier No. TRCG93151010MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 317318-69-7
Molecular Formula C22H20F3NO3S2
Quantity 10 mg
Synonym 2-[2-Methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]acetic Acid, Methyl-methyl-trifluoromethylphenyl -thiazolyl-methylsulfanyl-phenoxy-acetic Acid Methyl Ester, GW 1516 Methyl Ester
InChI Formula InChI=1S/C22H20F3NO3S2/c1-13-10-17(8-9-18(13)29-11-20(27)28-3)30-12-19-14(2)26-21(31-19)15-4-6-16(7-5-15)22(23,24)25/h4-10H,11-12H2,1-3H3
SMILES n1ccc(cc1)Nc2c(N)cccc2
Molecular Weight (g/mol) 467.52
Chemical Name or Material GW 501516 Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.