missing translation for 'onlineSavingsMsg'
Learn More

GW 501516, TRC

Product Code. 30012720
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012720 1 mg 1mg
30009414 25 mg 25mg
30009237 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012720 Supplier Toronto Research Chemicals Supplier No. TRCG9315001MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317318-70-0
Molecular Formula C21 H18 F3 N O3 S2
Molecular Weight (g/mol) 453.5
Synonym Acetic acid, 2-[2-methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]-, Acetic acid, [2-methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]- (9CI), 2-[2-Methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]acetic acid, 2-[2-Methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]thiazol-5-yl]methyl]thio]phenoxy]acetic acid, GW 1516, GW 501516, Cardarine, Endurobol, Acetic acid, 2-[2-methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]-, Acetic acid, [2-methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]- (9CI), 2-[2-Methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]acetic acid, 2-[2-Methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]thiazol-5-yl]methyl]thio]phenoxy]acetic acid, GW 1516, GW 501516, Cardarine, Endurobol
IUPAC Name 2-[2-methyl-4-[[4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl]methylsulfanyl]phenoxy]acetic acid
SMILES Cc1cc(SCc2sc(nc2C)c3ccc(cc3)C(F)(F)F)ccc1OCC(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H18F3NO3S2/c1-12-9-16(7-8-17(12)28-10-19(26)27)29-11-18-13(2)25-20(30-18)14-3-5-15(6-4-14)21(22,23)24/h3-9H,10-11H2,1-2H3,(H,26,27)
Formula Weight 453.068
Percent Purity >95%
Chemical Name or Material GW 501516
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.