missing translation for 'onlineSavingsMsg'
Learn More

GW 5074, MedChemExpress

Product Code. p-200099431
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480387

Brand: Medchem Express HY10542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW 5074 is a potent and selective c-Raf inhibitor with IC50 of 9 nM, and has no effect on the activities of JNK1/2/3, MEK1, MKK6/7, CDK1/2, c-Src, p38 MAP, VEGFR2 or c-Fms.

GW 5074 is a potent and selective c-Raf inhibitor with IC50 of 9 nM, and has no effect on the activities of JNK1/2/3, MEK1, MKK6/7, CDK1/2, c-Src, p38 MAP, VEGFR2 or c-Fms.

Specifications

Chemical Name or Material GW 5074
CAS 220904-83-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H8Br2INO2
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (191.96 mM)
SMILES O=C1NC2=C(/C1=C/C3=CC(Br)=C(C(Br)=C3)O)C=C(C=C2)I
Molecular Weight (g/mol) 520.94
Formula Weight 520.94
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.