missing translation for 'onlineSavingsMsg'
Learn More

GW0742, MedChemExpress

Product Code. 16465918
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465918 10 mg 10mg
16452067 50 mg 50mg
16428557 5 mg 5mg
16416366 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465918 Supplier Medchem Express Supplier No. HY13928
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GW0742 is a potent PPARβ and PPARδ agonist, with an IC50 of 1 nM for human PPARδ in binding assay, and EC50s of 1 nM, 1.1 μM and 2 μM for human PPARδ, PPARα, and PPARγ, respectively.

GW0742 is a potent PPARβ and PPARδ agonist, with an IC50 of 1 nM for human PPARδ in binding assay, and EC50s of 1 nM, 1.1 μM and 2 μM for human PPARδ, PPARα, and PPARγ, respectively.

Specifications

Chemical Name or Material GW0742
CAS 317318-84-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H17F4NO3S2
Quantity 10 mg
Synonym GW610742
Solubility Information DMSO : ≥ 34 mg/mL (72.11 mM)
SMILES O=C(O)COC1=CC=C(SCC2=C(C)N=C(C3=CC=C(C(F)(F)F)C(F)=C3)S2)C=C1C
Molecular Weight (g/mol) 471.49
Formula Weight 471.49
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.