missing translation for 'onlineSavingsMsg'
Learn More

GW2580, MedChemExpress

Product Code. 16471586
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471586 500 mg 500mg
16460617 50 mg 50mg
16454288 10 mg 10mg
16441006 200 mg 200mg
16425216 100 mg 100mg
16451129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16471586 Supplier Medchem Express Supplier No. HY10917
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GW2580 is an orally bioavailable and selective inhibitor of c-Fms kinase which completely inhibits human cFMS kinase in vitro at 0.06 μM. GW2580 acts as a competitive inhibitor of ATP binding to the cFMS kinase and inhibits colony-stimulating-factor-1 signaling.

GW2580 is an orally bioavailable and selective inhibitor of c-Fms kinase which completely inhibits human cFMS kinase in vitro at 0.06 μM. GW2580 acts as a competitive inhibitor of ATP binding to the cFMS kinase and inhibits colony-stimulating-factor-1 signaling.

Specifications

Chemical Name or Material GW2580
CAS 870483-87-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22N4O3
Quantity 500 mg
Solubility Information DMSO : 33.33 mg/mL (90.96 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (insoluble)
SMILES NC1=NC(N)=C(CC2=CC(OC)=C(OCC3=CC=C(OC)C=C3)C=C2)C=N1
Molecular Weight (g/mol) 366.41
Formula Weight 366.41
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.