missing translation for 'onlineSavingsMsg'
Learn More

GW3965 hydrochloride, MedChemExpress

Product Code. 16484078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484078 10 mg 10mg
16430437 50 mg 50mg
16415056 100 mg 100mg
16406787 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484078 Supplier Medchem Express Supplier No. HY10627A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.

GW3965 hydrochloride is a potent and selective liver X receptor (LXR) agonist with EC50s of 190 nM and 30 nM for hLXRα and hLXRβ, respectively.

Specifications

Chemical Name or Material GW3965 hydrochloride
CAS 405911-17-3
Color White
Purity Grade Notes Research
Molecular Formula C33H32Cl2F3NO3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (161.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES ClC1=C(C(F)(F)F)C=CC=C1CN(CC(C2=CC=CC=C2)C3=CC=CC=C3)CCCOC4=CC(CC(O)=O)=CC=C4.[H]Cl
Molecular Weight (g/mol) 618.51
Formula Weight 618.51
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.