missing translation for 'onlineSavingsMsg'
Learn More

GW6471, MedChemExpress

Product Code. 16499057
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499057 5 mg 5mg
16456746 100 mg 100mg
16442577 50 mg 50mg
16416478 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499057 Supplier Medchem Express Supplier No. HY15372

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW6471 is a potent PPARα antagonist.

GW6471 is a potent PPARα antagonist.

Specifications

Chemical Name or Material GW6471
CAS 880635-03-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C35H36F3N3O4
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (201.72 mM; Need ultrasonic)
SMILES CCC(NC[C@@H](N/C(C)=C\C(C1=CC=C(C(F)(F)F)C=C1)=O)CC2=CC=C(OCCC3=C(C)OC(C4=CC=CC=C4)=N3)C=C2)=O
Molecular Weight (g/mol) 619.67
Formula Weight 619.67
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.