missing translation for 'onlineSavingsMsg'
Learn More

GW9662, MedChemExpress

Product Code. 16497078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497078 10 mg 10mg
16493526 25 mg 25mg
16449627 5 mg 5mg
16447126 100 mg 100mg
16443127 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497078 Supplier Medchem Express Supplier No. HY16578
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GW9662 is a potent and selective PPARγ antagonist with an IC50 of 3.3 nM, showing 10 and 1000-fold selectivity over PPARα and PPARδ, respectively.

GW9662 is a potent and selective PPARγ antagonist with an IC50 of 3.3 nM, showing 10 and 1000-fold selectivity over PPARα and PPARδ, respectively.

Specifications

Chemical Name or Material GW9662
Physical Form Solid
CAS 22978-25-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H9ClN2O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (361.43 mM)
SMILES O=C(NC1=CC=CC=C1)C2=CC([N+]([O-])=O)=CC=C2Cl
Molecular Weight (g/mol) 276.68
Formula Weight 276.68
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.