missing translation for 'onlineSavingsMsg'
Learn More

H-1152 dihydrochloride, MedChemExpress

Product Code. 16459317
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459317 5 mg 5mg
16456768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16459317 Supplier Medchem Express Supplier No. HY15720A
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress H-1152 dihydrochloride is a membrane-permeable and selective ROCK inhibitor, with a Ki value of 1.6 nM, and an IC50 value of 12 nM for ROCK2.

H-1152 dihydrochloride is a membrane-permeable and selective ROCK inhibitor, with a Ki value of 1.6 nM, and an IC50 value of 12 nM for ROCK2.

Specifications

Chemical Name or Material H-1152 dihydrochloride
CAS 871543-07-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H23Cl2N3O2S
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (127.44 mM; Need ultrasonic) ∣H2O : 35.71 mg/mL (91.02 mM; Need ultrasonic)
SMILES CC1=CN=CC2=C1C(S(=O)(N3[C@@H](C)CNCCC3)=O)=CC=C2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 392.34
Formula Weight 392.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.