missing translation for 'onlineSavingsMsg'
Learn More

H-1152 Dihydrochloride, TRC

Product Code. 30403999
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403999 5 mg
30389108 10 mg
30402332 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403999 Supplier Toronto Research Chemicals Supplier No. TRCH0115205MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871543-07-6
Molecular Formula C16 H21 N3 O2 S . 2 Cl H
Molecular Weight (g/mol) 392.34
Synonym H 1152 dihydrochloride
IUPAC Name 4-methyl-5-[[(2S)-2-methyl-1,4-diazepan-1-yl]sulfonyl]isoquinoline;dihydrochloride
SMILES Cl.Cl.C[C@H]1CNCCCN1S(=O)(=O)c2cccc3cncc(C)c23

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H21N3O2S.2ClH/c1-12-9-18-11-14-5-3-6-15(16(12)14)22(20,21)19-8-4-7-17-10-13(19)2;;/h3,5-6,9,11,13,17H,4,7-8,10H2,1-2H3;2*1H/t13-;;/m0../s1
Formula Weight 391.0888
Percent Purity >95%
Chemical Name or Material H-1152, Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.