accessibility menu, dialog, popup

UserName

CAS RN 871543-07-6

CH3NHNSOONCH3HClHCl

CAS RN 871543-07-6

IUPAC Name: 4-methyl-5-[(2-methyl-1,4-diazepan-1-yl)sulfonyl]isoquinoline dihydrochloride
Synonyms: 4-methyl-5-(2-methyl-1,4-diazepan-1-ylsulfonyl)isoquinoline dihydrochloride
Molecular Weight (g/mol): 392.34
Molecular Formula: C16H23Cl2N3O2S
InChi Key: BFOPDSJOLUQULZ-UHFFFAOYNA-N
SMILES: Cl.Cl.CC1CNCCCN1S(=O)(=O)C1=C2C(C)=CN=CC2=CC=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results