Learn More
Invitrogen™ H3K27M Recombinant Rabbit Monoclonal Antibody (HL2794)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556740
Product Code. 30276193
  • 500.00 € / 100µL
Description
H3K27M Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
H3K27M
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P68431
HIST1H3A
Synthetic peptide of Histone H3 (K27M Mutant).
100 μL
Primary
Human
Antibody
IgG
Western Blot, Immunocytochemistry
HL2794
Unconjugated
HIST1H3A
H3FA; H3FB; H3FC; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; HIST1H3A; HIST1H3B; HIST1H3C; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J
Rabbit
Protein A
RUO
8350
-20°C, Avoid Freeze/Thaw Cycles