Learn More
Erlab H2O2 Bio-decontamination System, Droit
Product Code. 31794037 Shop All Erlab Products
Change view
Click to view available options
Material:
Epoxy-coated Steel
Unit Size:
Each
Product Code. Material Unit Size
31794037 Epoxy-coated Steel Each
1 options
Product Code. 31794037 Supplier Erlab Supplier No. NORT000071/D
  • ONLINE EXCLUSIVE PRICE
    3670.00 €
    3423.00 € / Pack of 1
    Save 247.00 €
    6% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hydrogen peroxide bio-decontamination technology can effectively eliminate even the most resistant microorganisms from the working volume of the chamber. A reliable, automated and validatable solution designed to meet the highest standards of hygiene, quality and safety.
  • Equipment and Options for Erlab Enclosures and Safety Cabinets: This equipment completes and adapts Erlab ductless enclosures and microbiological safety cabinets to the specific requirements of each laboratory. Every option is designed and validated by Erlab to be integrated without compromising the containment and protection performance of the equipment.
  • Designed and Validated by Erlab:
    • Components engineered for a specific enclosure or cabinet reference
    • Integration that preserves airflow, containment and operator protection
    • Materials selected for chemical and biological resistance and easy decontamination
    • Compatible with Erlab connected monitoring and safety programmes
  • Adapt Your Workstation to Your Protocols: Extraction options, front window and access solutions, decontamination systems, sealing accessories and internal fittings allow the workstation to be configured for the products handled, the equipment installed inside the enclosure and the ergonomics required by the operator.
  • Installation is designed to be carried out quickly, with no impact on the certification of the equipment when performed according to Erlab instructions.
  • Key Benefits:
    • Factory-validated option, fully compatible with the target equipment.
    • Maintains containment and operator protection performance.
    • Materials suitable for laboratory chemical and biological environments.
    • Improves ergonomics, versatility and ease of use.
    • Fast installation with no impact on equipment certification.
Specifications
Product Type Equipment Containment Enclosure
Material Epoxy-coated Steel