missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H60 Antibody (RM0248-11G35), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12428F
This item is not returnable.
View return policy
Description
H60 Monoclonal specifically detects H60 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| H60 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Rat | |
| Protein A or G purified | |
| Cancer | |
| 15101 | |
| Store at 4C in the dark. | |
| IgG2 |
| Flow Cytometry | |
| RM0248-11G35 | |
| Flow Cytometry | |
| H60a | |
| Mouse recombinant protein of H60. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction