missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HABP1/C1QBP/GC1q R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | HABP1/C1QBP/GC1q R |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-4 uL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645496
|
Novus Biologicals
NBP2-99898-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600579
|
Novus Biologicals
NBP2-99898-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HABP1/C1QBP/GC1q R Polyclonal antibody specifically detects HABP1/C1QBP/GC1q R in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| HABP1/C1QBP/GC1q R | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 708 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C1q globular domain-binding protein, C1qBP, complement component 1 Q subcomponent-binding protein, mitochondrial, complement component 1, q subcomponent binding protein, GC1QBP, gC1qR, gC1Q-R, GC1q-R protein, Glycoprotein gC1qBP, HABP1p33, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, p32, SF2P32, splicing factor SF2-associated protein | |
| Produced in rabbits immunized with purified, recombinant Human HABP1/C1QBP/GC1q R (Accession#: NP_001203.1; His75-Gln282) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title