Learn More
Gilac™ HACCP Food Contact Container
Product Code. 17948911
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17948911 White Each
17988651 Yellow Each
17978511 Blue Each
17968911 Red Each
17918701 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17948911 Supplier Gilac™ Supplier No. G613221
Add to basket
Add to basket
HACCP Food Contact Container is ideal for storage, food preparation, and transporting foodstuffs.
  • HACCP round container.
  • Food contact certified.
  • For storage, food preparation and transporting foodstuffs.
  • Highly durable.
  • Reinforced bottom with anti-wear skirt.
  • Reinforced handles to prevent distortion during carrying.
  • Optional, choice of two lids.
  • External Diameter includes handles.
  • Width corresponds to diameter without handles.
Specifications
Product Type HACCP Food Container
Material Polyethylene
Color White
Volume (English) 13.2 gal.
Volume (Metric) 50 L
Product Title
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.