missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD23A Recombinant Rabbit Monoclonal Antibody (24GB1080)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557672
This item is not returnable.
View return policy
Description
RAD23A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.
Specifications
| RAD23A | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RAD23A | |
| 2310040P19Rik; AL024030; HHR23A; HR23A; MGC111083; Mhr23a; rad23; RAD23 homolog A (S. cerevisiae); RAD23 homolog A, nucleotide excision repair protein; RAD23, yeast homolog, A; RAD23A; RAD23a homolog (S. cerevisiae); UV excision repair protein RAD23 homolog A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19358, 5886 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1080 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P54725, P54726 | |
| RAD23A | |
| A synthesized peptide derived from human RAD23A (aa 205-250). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction