missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HADHA Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human HADHA. Source: E.coli Amino Acid Sequence: DEDIQFRLVTRFVNEAVMCLQEGILATPAEGDIGAVFGLGFPPCLGGPFRFVDLYGAQKIVDRLKKYEAAYGKQFTPCQLLADHANSPNKK The HADHA Recombinant Protein Antigen is derived from E. coli. The HADHA Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Purity | Greater than 80% by SDS-PAGE and Coomassie blue staining |
| Common Name | HADHA Recombinant Protein Antigen |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Antibody Competition |
| Molecular Weight (g/mol) | 28 kDa |
| Product Type | Recombinant Protein Antigen |
| Quantity | 100 μL |
| Sequence | DEDIQFRLVTRFVNEAVMCLQEGILATPAEGDIGAVFGLGFPPCLGGPFRFVDLYGAQKIVDRLKKYEAAYGKQFTPCQLLADHANSPNKK |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-50314. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction