missing translation for 'onlineSavingsMsg'
Learn More

Hafnium Standard: Hf @ 1000 μg/mL in 5% HCl, VHG™

Product Code. 17994336
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994336 50 mL 50mL
17919009 100 mL 100mL
17943556 250 mL 250mL
17940426 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17994336 Supplier VHG Labs Supplier No. VHGPHFH50
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Hafnium Standard: Hf @ 1000 μg/mL in 5% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hafnium
CAS 7440-58-6
Methods ICP/ICP-MS
Analyte or Component Names Hafnium
Shipping Condition Room Temperature
Molecular Formula Hf
Certifications/Compliance ISO 17034
Synonym Hf
InChI Formula InChI=1S/Hf
SMILES [Hf]
IUPAC Name hafnium
Solvent or Matrix Dilute HCl
Molecular Weight (g/mol) 178.49
Quantity 50 mL
Formula Weight 179.947 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.