missing translation for 'onlineSavingsMsg'
Learn More

Hafnium Standard: Hf @ 1000 ug/mL in 2% HNO3/0.5%HF

Product Code. 30318842
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318842 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318842 Supplier VHG Labs Supplier No. VHGPHFNF100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hafnium
CAS 7440-58-6
Analyte or Component Names Hafnium
Shipping Condition Room Temperature
Molecular Formula Hf
Concentration 1000 μg/mL
Synonym Hf
InChI Formula InChI=1 S/Hf
Solution Type Single Solution
SMILES [Hf]
Element Hf
IUPAC Name hafnium
Solvent or Matrix Dilute HNO3
Industry Type Food Testing
Molecular Weight (g/mol) 178.49
Quantity 100 mL
Formula Weight 179.947
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.