missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HAGH Polyclonal Antibody

Product Code. 30534717
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30534717 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534717 Supplier Invitrogen™ Supplier No. PA5149101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

The enzyme encoded by the HAGH gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutathione and D-lactate. Three transcript variants encoding different isoforms have been found for this gene. Diseases associated with HAGH include Hydroxyacyl Glutathione Hydrolase Deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAGH
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene HAGH
Gene Accession No. O35952, Q16775, Q99KB8
Gene Alias BC019817; GLO2; Glo-2; Glx II; GLX2; GLXII; Glyoxalase II; glyoxylase 2; HAGH; HAGH1; hydroxyacyl glutathione hydrolase; hydroxyacylglutathione hydrolase; hydroxyacylglutathione hydrolase, mitochondrial; hydroxyacylglutathione hydroxylase; round spermatid protein RSP29; Rsp29
Gene Symbols HAGH
Host Species Goat
Immunogen E. coli-derived recombinant human Glyoxalase II, Met49-Ile254.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14651, 24439, 3029
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.