missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAI-1/HGFA Inhibitor 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89720
This item is not returnable.
View return policy
Description
HAI-1/HGFA Inhibitor 1 Polyclonal antibody specifically detects HAI-1/HGFA Inhibitor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| HAI-1/HGFA Inhibitor 1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20-1:50 | |
| HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ACFLINCLYEQNFVCKFAPREGFINYLTREVYRSYRQLRTQGFGGSGIPKAWAGIDLKVQPQEPLVLKDVENTDWRLLRGDTDVRVERKDPNQVELWGLKEGTYLFQLTVTSSDHP | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6692 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction