missing translation for 'onlineSavingsMsg'
Learn More

Halofuginone hydrobromide, MedChemExpress

Product Code. 16483926
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483926 25 mg 25mg
16449238 10 mg 10mg
16401658 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16483926 Supplier Medchem Express Supplier No. HYN1584A
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Halofuginone (RU-19110) hydrobromid, a Febrifugine derivative, is a competitive prolyl-tRNA synthetase inhibitor with a Ki of 18.3 nM.

Halofuginone hydrobromid is a specific inhibitor of type-I collagen synthesis and attenuates osteoarthritis (OA) by inhibition of TGF-β activity. Halofuginone hydrobromid is also a potent pulmonary vasodilator by activating Kv channels and blocking voltage-gated, receptor-operated and store-operated Ca2+ channels. Halofuginone hydrobromid has anti-malaria, anti-inflammatory, anti-cancer, anti-fibrosis effects.

Specifications

Chemical Name or Material Halofuginone hydrobromide
Physical Form Solid
CAS 64924-67-0
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C16H18Br2ClN3O3
For Use With (Application) COVID-19-anti-virus
Synonym RU-19110 hydrobromide
Solubility Information DMSO : 50 mg/mL (100.89 mM; Need ultrasonic) ∣H2O : 2.6 mg/mL (5.25 mM; Need ultrasonic)
SMILES O=C1N(CC(C[C@@H]2NCCC[C@H]2O)=O)C=NC3=C1C=C(Cl)C(Br)=C3.Br
Molecular Weight (g/mol) 495.59
Formula Weight 495.59
Percent Purity 99.55%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.